missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ PYROGENT™-5000 Kinetic Turbidimetric LAL Assay

Quantitative, kinetic assay for the detection of Gram-negative bacterial endotoxin.

Brand:  Lonza™ N384

Product Code. 11650321

  • £1190.00 / 200 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Quantitative, kinetic assay.
  • Bulk turbidimetric kinetic LAL Assay for endotoxin testing for 200 tests.
  • Kit contains endotoxin and reconstitution buffer.
  • Detects Gram-negative bacterial endotoxin.
  • Sensitivity range from 0.01 to 100 EU/mL.
  • Cost-effective method for water and large-volume parenterals testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Turbidimetric kinetic LAL assay
200 tests
Endotoxin testing
2 × 100-test vials of Lysate, 2 viaLs reconstitution buffer, 1 vial endotoxin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers