missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole, 99%, extra pure

Product Code. 10498310 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10498310

Brand: Thermo Scientific Acros 157711000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-97-7 | C4H5N | 67.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-97-7
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.09
MDL Number MFCD00005216
InChI Key KAESVJOAVNADME-UHFFFAOYSA-N
Synonym pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
PubChem CID 8027
ChEBI CHEBI:19203
IUPAC Name 1H-pyrrole
SMILES N1C=CC=C1

Specifications

Melting Point -23.0°C
Color Brown to Colorless
Density 0.9600g/mL
Boiling Point 129.0°C to 131.0°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5080 to 1.5100
Quantity 100 g
Beilstein 20,159
Merck Index 15,8128
Specific Gravity 0.96
Solubility Information Solubility in water: 60g/L (20°C). Other solubilities: insoluble in dilute alkalies, soluble in alcohol, ether and dilute acids, soluble in most organic solvents
Viscosity 1.317 mPa.s (20°C)
Formula Weight 67.09
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Pyrrole
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pyrrole
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.