missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine, 99%

Product Code. 11412637
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11412637

Brand: Thermo Scientific Alfa Aesar A14852.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-75-1 | C4H10ClN | 107.58 g/mol

Pyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.

Solubility
Miscible with alcohol, ether, chloroform and water.

Notes
Store in cool place. Incompatible with acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-75-1
Molecular Formula C4H10ClN
Molecular Weight (g/mol) 107.58
MDL Number MFCD00005249
InChI Key FCLZCOCSZQNREK-UHFFFAOYSA-N
Synonym tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
PubChem CID 31268
ChEBI CHEBI:33135
IUPAC Name hydrogen pyrrolidine chloride
SMILES [H+].[Cl-].C1CCNC1

Specifications

Melting Point -63°C
Density 0.856
Boiling Point 86°C to 88°C
Flash Point 2°C (35°F)
Assay Percent Range 99%
Odor Like Rotten Eggs
Refractive Index 1.4431
Quantity 100 mL
UN Number UN1922
Beilstein 102395
Sensitivity Air Sensitive
Merck Index 14,8015
Solubility Information Miscible with alcohol,ether,chloroform and water.
Formula Weight 71.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Pyrrolidine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pyrrolidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Pyrrolidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.