missing translation for 'onlineSavingsMsg'
Learn More
Pyrrolidine, 99+%, Thermo Scientific Chemicals Product Code.: 10163710

Pyrrolidine, 99+%, Thermo Scientific Chemicals

Product Code. 10163710
250 mL, Glass bottle
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10163710

Brand: Thermo Scientific Acros 132082500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-75-1
Molecular Formula C4H10ClN
Molecular Weight (g/mol) 107.58
MDL Number MFCD00005249
InChI Key FCLZCOCSZQNREK-UHFFFAOYSA-N
Synonym tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
PubChem CID 31268
ChEBI CHEBI:33135
IUPAC Name hydrogen pyrrolidine chloride
SMILES [H+].[Cl-].C1CCNC1

Specifications

Melting Point -63.0°C
Density 0.8660g/mL
Boiling Point 86.0°C to 88.0°C
Flash Point 3°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4421 to 1.4441
Quantity 250 mL
Beilstein 20,4
Fieser 01,972; 02,354; 03,240; 07,309; 08,428; 13,14
Merck Index 15,8129
Specific Gravity 0.866
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether and chloroform, soluble in most organic solvents
Viscosity 0.94 mPa.s (20°C)
Formula Weight 71.11
Percent Purity 99+%
Chemical Name or Material Pyrrolidine, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468573 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number132081000,132080010,132080100,132082500,132080025
Lot NumberA0462838 Certificate TypeCertificate of Analysis Date22/05/2024 Catalog Number132080050,132080010,132081000,132080100,132082500,132080025
Lot NumberA0444022 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number132080050,132080010,132081000,132080100,132082500,132080025
Lot NumberA0458434 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog Number132080050,132080010,132081000,132080100,132082500,132080025
Lot NumberA0432003 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number132080050,132080010,132081000,132080100,132082500,132080025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pyrrolidine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrrolidine, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.