missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic acid, sodium salt, 99+%, Thermo Scientific Chemicals

£27.24 - £288.44

Chemical Identifiers

CAS 113-24-6
Molecular Formula C3H3NaO3
Molecular Weight (g/mol) 110.04
MDL Number MFCD00002586
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Synonym sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
PubChem CID 23662274
ChEBI CHEBI:50144
IUPAC Name sodium;2-oxopropanoate
SMILES CC(=O)C(=O)[O-].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10448100
View Documents
Thermo Scientific Acros
132150250
25 g Plastic bottle
£27.24
25g
Please to purchase this item. Need a web account? Register with us today!
10376680
View Documents
Thermo Scientific Acros
132151000
100 g Plastic bottle
£73.26
100g
Please to purchase this item. Need a web account? Register with us today!
10328703
View Documents
Thermo Scientific Acros
132155000
500 g Plastic bottle
£288.44
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

113-24-6
110.04
DAEPDZWVDSPTHF-UHFFFAOYSA-M
23662274
sodium;2-oxopropanoate
C3H3NaO3
MFCD00002586
sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
CHEBI:50144
CC(=O)C(=O)[O-].[Na+]

Specifications

113-24-6
Colorless to Yellow
0.5% max.
99+%
CH3COCO2Na
03, 608
(10% in water) Clear colorless to light yellow
CC(=O)C(=O)[O-].[Na+]
110.04
CHEBI:50144
99+%
Pyruvic acid, sodium salt
>300.0°C
5.5 to 6.5 (10% soln. at 20°C)
Authentic
C3H3NaO3
MFCD00002586
sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
DAEPDZWVDSPTHF-UHFFFAOYSA-M
sodium;2-oxopropanoate
23662274
110.04
Solution
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

missing translation for 'einecsNumber' : 204-024-4

Product Identifier
  • Pyruvic acid, sodium salt

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.