missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ pZerve Cryopreservation Solution

Product Code. p-8015224 Shop All MP Biomedicals Products
Click to view available options
Quantity:
20 mL
60 mL
Unit Size:
20mL
60mL
This item is not returnable. View return policy

Product Code. 15757879

Brand: MP Biomedicals™ 0920303M2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cryopreservation solution that does not contain dimethyl sulfoxide (DMSO) and serum.

pZerve has been successfully used with a variety of cell types including mast cells, dendritic cells, and mesenchymal cells. It can be used for cells cultured in serum-free medium or with serum-containing medium.

TRUSTED_SUSTAINABILITY

Specifications

Form Solution
Product Type Cryopreservation Media
For Use With (Application) Cryopreservation
Quantity 60 mL
Content And Storage Store at 2° to 8°C
Description pZerve Cryopreservation Media
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.