missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ Q-CLASSIC-30 One-Door Light Gray Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas with fire resistance up to 30 minutes.
Brand: Asecos™ 3100806133627
This item is not returnable.
View return policy
Description
- The sophisticated design and use of melamine-resin-coated, fireproof special plates ensures an optimum price/performance ratio
- Integrated transport base for simple in-house transport, can be transported upright through doors
- Adjustable feet installed in the base allow the safe and fast compensation of minor surface unevenness
- Modern bicolor design and language-free labelling for optimum international usability
- Door lockable with cylinder lock, and can be integrated by the customer in an existing locking system
- Interior equipment such as drawers, height-adjustable shelves and bottom collecting sumps available
- Fire resistant up to 30 minutes
- Triple-hinged doors, stoppable in any position, self-closing in the event of fire
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system for ventilation
- In accordance with EN 14470-1 and TRGS 510
Specifications
EN 14470-1, TRGS 510 | |
519 x 750 x 1626 mm | |
Wing | |
Drawers: 60 kg/19 L each, Maximum load: 600 kg | |
Plastic laminated | |
Light Gray | |
Safety storage cabinet |
4 x 60 kg/19 L sheet steel drawers | |
620 x 864 x 1947 mm | |
Storage of flammable and hazardous materials | |
1 | |
253 kg | |
Melamine Resin |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Asecos™ Q-CLASSIC-30 One-Door Light Gray Safety Storage Cabinets > 4x 60 kg/19 L sheet steel drawers
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur