Learn More
Asecos™ Q-CLASSIC-30 Two-Door Yellow Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas with fire resistance up to 30 minutes.
Brand: Asecos™ 3100306033649
2647.70 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 222.00000kg
Description
- The sophisticated design and use of melamine-resin-coated, fireproof special plates ensures an optimum price/performance ratio
- Integrated transport base for simple in-house transport, can be transported upright through doors
- Adjustable feet installed in the base allow the safe and fast compensation of minor surface unevenness
- Modern bicolor design and language-free labelling for optimum international usability
- Door lockable with cylinder lock, and can be integrated by the customer in an existing locking system
- Interior equipment such as drawers, height-adjustable shelves and bottom collecting sumps available
- Fire resistant up to 30 minutes
- Triple-hinged doors, stoppable in any position, self-closing in the event of fire
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system for ventilation
- In accordance with EN 14470-1 and TRGS 510
Specifications
6 x 60 kg/29 L sheet steel drawers | |
Storage of flammable and hazardous materials | |
Melamine Resin | |
334 kg | |
Yellow | |
Wing | |
EN 14470-1, TRGS 510 |
Safety storage cabinet | |
Plastic laminated | |
620 x 1164 x 1947 mm | |
519 x 1050 x 1626 mm | |
6 sheet steel drawers: 60 kg/29 L each, Maximum load: 600 kg | |
2 |