Learn More
asecos™ Type 90 Safety Storage Cabinet Q-CLASSIC-90
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3004104033622
Alert:
For indoor use only
Description
Safe and approved storage of flammable hazardous substances in working areas
Function / construction
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
Certificates
- Fire resistance 90 minutes
- type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
Fire resistance 90 minutes; type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
615 mm | |
Wing doors in warning yellow | |
615 x 893 x 1953 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
1647 mm | |
2 | |
Standard | |
395 kg | |
750 mm | |
Stainless Steel |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | |
522 mm | |
522 x 750 x 1647 mm | |
Wing | |
1953 mm | |
Q90.195.090 | |
3 | |
Powder-coated textured | |
893 mm | |
Anthracite Gray, Warning Yellow | |
Safety storage cabinet |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.