missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ Q-CLASSIC-90 Two-Door Light Gray Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas.
POA
Specifications
Certifications/Compliance | EN 14470-1 |
---|---|
Color | Light Gray |
Product Type | Flammables Storage Cabinet |
Door Style | Triple-hinged |
For Use With (Application) | Storage of flammable hazardous substances |
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
15860448
|
Asecos™
3000104133635 |
75 kg Shelves, 75 kg Insert, 33 L Sump |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15870448
|
Asecos™
3000104133647 |
4 x 60 kg/29 L Drawers |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15880448
|
Asecos™
3004104133627 |
4 x 60 kg/19 L Drawers |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Extremely robust design with scratch-proof surface
- Triple-hinged doors
- Safety elements assembled outside the storage compartment for increased protection against corrosion
- Integrated transport base for simple in-house transport
- Cabinet can be transported upright through doors
- Adjustable feet in base allow the safe and fast compensation of minor surface unevenness
- Modern bicolor design - choice of seven different door colors
- Language-free labelling for optimum international usability
- Doors lockable with cylinder lock, cabinets can be integrated by the customer in an existing master key system
- Locking status indicator (red/green) signals the locking status
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Visual inspection of the exhaust air function in the exhaust grille (rotating fan impeller)
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- In accordance with EN 14470-1 and TRGS 510
- Fire resistance of 90 minutes
Specifications
EN 14470-1 | |
Flammables Storage Cabinet | |
Storage of flammable hazardous substances |
Light Gray | |
Triple-hinged | |
2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Asecos™ Q-CLASSIC-90 Two-Door Light Gray Safety Storage Cabinets