missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ Q-CLASSIC-90 Two-Door Yellow Safety Storage Cabinets

For the safe and approved storage of flammable hazardous substances in working areas.

POA

Specifications

Certifications/Compliance EN 14470-1
Color Yellow
Product Type Safety storage cabinet
Door Style Triple-hinged
For Use With (Application) Storage of flammable hazardous substances
View More Specs

Products 4
Product Code Brand Load Bearing Capacity Price Quantity & Availability  
Product Code Brand Load Bearing Capacity Price Quantity & Availability  
15768362
View accessory products View Documents
Asecos™
3000104033647
4 x Drawers 60 kg/29 L each
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15771423
View accessory products View Documents
Asecos™
3000104033649
6 x Drawers 60 kg/29 L each
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15825111
View Documents
Asecos™
34632-040-34637
3 x Shelves 75 kg each; Insert 75 kg; Sump 33 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15983690
View accessory products View Documents
Asecos™
3004104033629
6 x Drawers 60 kg/19 L each
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

  • Extremely robust design with scratch-proof surface
  • Triple-hinged doors
  • Safety elements assembled outside the storage compartment for increased protection against corrosion
  • Integrated transport base for simple in-house transport
  • Cabinet can be transported upright through doors
  • Adjustable feet in base allow the safe and fast compensation of minor surface unevenness
  • Modern bicolor design - choice of seven different door colors
  • Language-free labelling for optimum international usability
  • Doors lockable with cylinder lock, cabinets can be integrated by the customer in an existing master key system
  • Locking status indicator (red/green) signals the locking status
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • Visual inspection of the exhaust air function in the exhaust grille (rotating fan impeller)
  • Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
  • In accordance with EN 14470-1 and TRGS 510
  • Fire resistance of 90 minutes
Specifications

Specifications

EN 14470-1
Safety storage cabinet
Storage of flammable hazardous substances
Yellow
Triple-hinged
2
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Asecos™ Q-CLASSIC-90 Two-Door Yellow Safety Storage Cabinets

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.