missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ Q-PEGASUS-90 Two-Door Yellow Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas.
£3497.47 - £4582.38
Specifications
Certifications/Compliance | EN 14470-1 |
---|---|
Color | Yellow |
Dimensions (D x W x H) | 615 x 893 x 1953 mm |
Product Type | Safety storage cabinet |
Door Style | Auto-close |
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Load Bearing Capacity | Price | Quantity & Availability | |||||
15759428
|
Asecos™
3004204033629 |
Drawers 60 kg/19 L each |
£4582.38
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15871408
|
Asecos™
3004204033622 |
Shelves 75 kg; Insert 75 kg; Sump 22 L |
£3497.47
Each |
Estimated Shipment: 05-12-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Extremely robust design with scratch-proof surface
- Triple-hinged doors
- Safety elements assembled outside the storage compartment for increased protection against corrosion
- One-hand operation for double-door cabinets
- Container remains in one hand whilst opening
- Doors remain open in the serial door open arrest system, convenient and safe removal or storage of containers
- Doors automatically close after approx. 60 seconds
- Acoustic and visual signal before door closing
- Modern bicolor design - choice of seven different door colors
- Language-free labelling for optimum international usability
- Doors lockable with cylinder lock, cabinets can be integrated by the customer in an existing master key system
- Locking status indicator (red/green) signals the locking status
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Visual inspection of the exhaust air function in the exhaust grille (rotating fan impeller)
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- In accordance with EN 14470-1 and TRGS 510
- Fire resistance of 90 minutes
- Integrated transport base with removable, optional base cover
- Easy alignment – adjusting aids to compensate for uneven floors
Specifications
EN 14470-1 | |
615 x 893 x 1953 mm | |
Auto-close | |
2 |
Yellow | |
Safety storage cabinet | |
Storage of flammable hazardous substances |