missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Qdot™ 605 Biotin Conjugate Kit
Qdot™ 605 Biotin Conjugate Kit
Brand: Molecular Probes™ Q10301MP
Code : 50
Additional Details : Weight : 0.00450kg
Description
The biotin-labeled Qdot™ 605 nanocrystals are available for detecting streptavidin probes or for creating noncovalent conjugates with streptavidin-labeled molecules or with other biotinylated molecules using a streptavidin bridge. The product is provided as 250μL of a 2μM solution and includes 30 mL of Qdot™ incubation buffer.Specifications
1 kit | |
Room Temperature | |
Biotin Qdot Conjugation Kit |
Store in refrigerator (2–8°C). | |
Qdot™ |
For Research Use Only. Not for use in diagnostic procedures.