Learn More
Swann-Morton™ Qlicksmart SnapIT Personal, Regular, Blue Ampule Opener
Safer way to open glass ampules and avoid painful and dangerous sharps injuries.
Brand: Swann-Morton™ SN-01R
This item is not returnable.
View return policy
Description
- This easy-to-use multi-use ampule opener helps to avoid unacceptable sharps injuries rates seen when ampules are opened by hand.
- Opening ampules is a particularly high-risk event with 26% of needlestick and sharps injuries (NSIs) being caused by opening an ampule or by broken ampules.
- The device was invented by a registered nurse in Rockhampton, Qld, Australia who recognized that millions of tamperproof glass ampules are used each year within health care and related practice to contain pharmaceuticals.
- The practical benefits are clear however the injury rates have always been significant.
- You simply insert the ampule lid into the multi use SnapIT making sure you have the correct size.
- If there is a dot on the ampule then align this with the dot on the SnapIT.
- Snap away from your body to open the ampule.
- You can then eject the glass ampule lid directly into a sharps container.
- There are both personal and trolley versions available in the range:
- Trolley Edition: for team use featuring a flat side so it can lay on a bench top or trolley without rolling away - perfect for hospital medication preparation areas.
- Pocket Edition: anodized aluminum for personal use on a key ring, hung from a lanyard or kept in your pocket.
- Lite Edition: made from sturdy, low cost plastic.
- Regular size Snap-It can be used with ampules of 1-2 mL, 5-10 mL and 10-15 mL and the large trolley edition can be used with ampules of 5-10 mL and 20-25 mL.
Specifications
Blue | |
Ampule Opener | |
Anodized Aluminum | |
Regular |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Swann-Morton™ Qlicksmart SnapIT Personal, Regular, Blue Ampule Opener > Personal, regular-size, blue
Spot an opportunity for improvement?Share a Content Correction