missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays
Utilize the precision, consistency and optimized performance of Thermo Scientific™ QMS™ TDM Assays, ready-to-use, liquid immunoassays.
£777.10
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Sample Type | Plasma, Serum |
Type | Reagent |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
17222484
|
Thermo Scientific™
0373589 |
Vancomycin Assay |
£777.10
Each |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Accurately measure therapeutic drug levels in serum and plasma with the Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays. These liquid, ready-to-use assays are a convenient alternative to more expensive and time-consuming technologies. QMS (Quantitative Microsphere System) immunoassays have excellent precision and accuracy, optimized performance on a wide range of general chemistry analyzers and lot-to-lot consistency.
Reduces Labor- Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
- Based on competitive inhibition immunoassay principle whereby free-drug in the sample competes for antibody binding sites with drug molecules coated onto uniform microparticles
- Liquid stable and ready-to-use
- Applicable to general chemistry analyzers
- Validated parameters assure lab-to-lab reproducibility
- Superb reagent stability
- Excellent correlation to reference methods
- Optimal sensitivity, precision and dynamic range
- Low cross-reactivity with metabolites and commonly co-administered drugs
- Low interference from endogenous substances
Specifications
2°C to 8°C | |
Plasma, Serum | |
Reagent |