Learn More
Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators
Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ TDM Calibrators, offered in Levels A to F.
£308.00
Product Code | Brand | Detectable Analytes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Detectable Analytes | Price | Quantity & Availability | ||||||
17232484
|
Thermo Scientific™
0373597 |
Vancomycin |
£308.00
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
Description
Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators, which employ advanced microparticle technology that is easy-to-use.
Reduces Labor- Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
- Based on competitive inhibition immunoassay principle whereby free-drug in the sample competes for antibody binding sites with drug molecules coated onto uniform microparticles
- Liquid stable and ready-to-use
- Applicable to general chemistry analyzers
- Validated parameters assure lab-to-lab reproducibility
- Superb reagent stability
- Excellent correlation to reference methods
- Low cross-reactivity with metabolites and commonly co-administered drugs
Specifications
2°C to 8°C | |
Plasma, Serum |