missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Qorpak™ Glass Bottle Beakers with Green Thermoset F217 and PTFE Caps
Click to view available options
Capacity:
8 oz., 240 mL
Height (Metric):
127 mm
Height (English):
5 in.
Quantity:
24/Cs.
Description
- Designed with slight shoulder for easy pouring and handling
- Molded graduations in mL and ounces
- Made of clear glass
- For use with biological and pathological specimens
- For storage of industrial laboratory chemicals and reagents
Specifications
Specifications
Capacity | 8 oz., 240 mL |
Capacity (Metric) | 240 mL |
Capacity (English) | 8 oz. |
Color | Clear |
Diameter (English) | 2.44 in. |
Material | Glass |
Quantity | 24/Cs. |
Closure Color | Green |
Height (Metric) | 127 mm |
Height (English) | 5 in. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'