Learn More
STEMCELL Technologies qPCR Master Mix Kit
For probe-based assays and arrays
Brand:  STEMCELL Technologies 07516
Code : Z1
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 16426800
  • £153.90 / Each
Estimated Shipment: 20-09-2024
to see stock.
Description
Description
  • qPCR Master Mix is a 2X concentrated solution optimized for TaqMan™ probe-based, real-time quantitative polymerase chain reaction (qPCR)
  • It is intended for use in combination with gene-specific primers (to amplify target cDNA) and fluorogenic-labeled probes that use the 5' nuclease activity of Taq DNA polymerase to produce a fluorescent signal
  • The rate of accumulation of the fluorescent signal is used to quantify the cDNA, and thereby to determine the amount of mRNA present in the original sample
  • qPCR Master Mix includes a Hot Start DNA polymerase, dNTPs, MgCl2, enhancers, and stabilizers
  • The qPCR Master Mix Kit also includes ROX Reference Dye as a separate component, making it compatible with both reference dye-dependent and -independent qPCR systems
  • Efficient, sensitive, and reproducible
  • Optimal performance when using standard or fast cycling conditions
  • Ideal for high-throughput applications and overnight experiments
  • Compatible with various real-time qPCR instruments
Specifications
Specifications
Adipocytes; Chondrocytes; Endoderm, PSC-Derived; Hepatic Cells; Intestinal Cells; Mesenchymal Stem and Progenitor Cells; Osteoblasts; Pancreatic Cells; Pluripotent Stem Cells
1 mL Kit
Real Time PCR (qPCR)