Learn More
Invitrogen™ Qualification Add-on For CL1000 Imaging System
Designed to help balance business and laboratory requirements and help comply with global standards, country-specific regulations, and other laboratory site requirements.
Brand: Invitrogen™ ZGQUSCIBRIGHTCL
This item is not returnable.
View return policy
Description
- Verify instrument performance and minimize downtime with IQ/OQ services (Installation Qualification and Operation Qualification)
- Complete with audit ready documentation, factory-trained and experienced engineers will conduct IQ/OQ services
- Qualification services help ensure instruments are installed and operating according to manufacturer’s specifications
- Helps move into production faster
- IQ/OQ recommended: At installation , after a move
- OQ recommended: After planned maintenance, After major repair, After additions, changes, or upgrades to the system, For a scheduled OQ site requirement
Specifications
iBright FL1000 Imaging System | |
Qualification Add-on | |
Warranty for Western Blotting instrumentation | |
Audit-ready documentation, Order and system verification, Documentation verification, Installation verification, Software verification, Hardware operation verification, Component verification, Service coverage verification, Customer training verification |