Learn More
BINDER™ Qualification Documents IQ/OQ
Qualification documents - IQ/OQ documents – supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature, humidity, and light values
Brand: BINDER™ 7057-0003
Additional Details : Weight : 0.00010kg
Description
- Parameters depending on unit
- Available as hardcopy inside folder or PDF document
Specifications
BINDER Chambers Series KBF P, KBF LQC, KBW, KBWF | |
On request | |
Supporting documents for validation performed by customers |
Qualification documents - Digital in PDF format, IQ/OQ documents - supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature, humidity, and light values | |
IQ/OQ Documentation (PDF) | |
IQ/OQ checklists include calibration guide and comprehensive unit documentation |