Learn More
BINDER™ Qualification Documents IQ/OQ
Qualification documents - IQ/OQ documents – supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature, humidity, and light values
Brand:  BINDER™ 7007-0003
Additional Details : Weight : 2.20000kg
 View more versions of this product
Product Code. 16434009
  • £1465.00 / Each
Estimated Shipment: 28-08-2024
to see stock.
Description
Description
  • Parameters depending on unit
  • Available as hardcopy inside folder or PDF document
Specifications
Specifications
BINDER Chambers Series KBF P, KBF LQC, KBW, KBWF
On request
Supporting documents for validation performed by customers
Qualification documents - Hard copy inside folder, IQ/OQ documents - supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature, humidity, and light values
IQ/OQ Documentation (Hardcopy)
IQ/OQ checklists include calibration guide and comprehensive unit documentation