Learn More
Macherey-Nagel™ Qualitative Analysis Test Papers, MN 615
Widely used for routine separations.
Brand: Macherey-Nagel™ 4310082
This item is not returnable.
View return policy
Description
- Qualitative filter papers widely used for routine separations
- Manufactured from top quality raw materials using ultra pure water to ensure consistent and reliable filtration
- Most widely used general purpose filter paper with medium retention and flow rate
- This filter paper covers a wide range of laboratory applications and is frequently used for clarifying liquids
Specifications
Disc | |
MN 615 | |
100/Pk. | |
82 mm |
Product Content Correction
Product Title
Macherey-Nagel™ Qualitative Analysis Test Papers, MN 615 > 82 mm diameter
Spot an opportunity for improvement?Share a Content Correction