Learn More
Macherey-Nagel™ Qualitative Analysis Test Papers, MN 615
Widely used for routine separations.
Brand:  Macherey-Nagel™ 4310025
 View more versions of this product
Product Code. 10499634
  • £5.70 / Pack of 100
Explore more special offers
Description
Description
  • Qualitative filter papers widely used for routine separations
  • Manufactured from top quality raw materials using ultra pure water to ensure consistent and reliable filtration
  • Most widely used general purpose filter paper with medium retention and flow rate
  • This filter paper covers a wide range of laboratory applications and is frequently used for clarifying liquids
Specifications
Specifications
Disc
MN 615
100/Pk.
25 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Qualitative Analysis Test Papers, MN 615 > 25 mm diameter