Learn More
Socorex™ Qualitix™ Micropipette Tips, 200μL
Superior, autoclavable micropipette tips cover a range of sizes and fit a broad array of micropipette brands. Socorex™ Qualitix™ Micropipette Tips, 200μL also come in a variety of shapes to facilitate fluid transfer.
Brand: Socorex™ 3070.0200B
Additional Details : Weight : 0.48000kg
Description
- Cover a range of sizes.
- Autoclavable.
- Optimal tightness fitting on Socorex™ pipettes and broad compatibility with other brands.
- Various tip versions available.
Specifications
200 μL | |
Non-filtered | |
Natural | |
Beveled | |
1/PK, 10PK/CS | |
Universal Fit Micro |
200μL | |
Non-sterile | |
Autoclavable | |
1000/Bag | |
Single-use | |
Qualitix |