Learn More
Sampling Systems™ Quality Control PharmaLabels
Medium strength adhesion paper labels with a special FDA acceptable adhesive. Sampling Systems™ Quality Control PharmaLabels are ideal for applications where the adhesive might come into contact with the product.
Brand:  Sampling Systems™ 8220P-031
 View more versions of this product
Product Code. 15819501
  • £67.65 / Pack of 250
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Medium strength adhesive
  • Large writing area
  • Each roll individually packed
  • FDA acceptable adhesive (CFR 175.105)
  • Ideal for applications where the adhesive might come into contact with the product
Specifications
Specifications
CFR 175.105
Blue
PharmaLabel
250/Pk.
150 x 150 mm
Food Grade Adhesive
Paper
Sampled
-40°C to +70°C