Learn More
Quality Control Standards 21 For ICP, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep QC-21-250
Code : NEW
Additional Details : CAS Number : 14797-71-8 Weight : 0.32000kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
As | 100μg/mL |
Be | 100μg/mL |
Ca | 100μg/mL |
Cd | 100μg/mL |
Co | 100μg/mL |
Cr | 100μg/mL |
Cu | 100μg/mL |
Fe | 100μg/mL |
Li | 100μg/mL |
Mg | 100μg/mL |
Mn | 100μg/mL |
Mo | 100μg/mL |
Ni | 100μg/mL |
Pb | 100μg/mL |
Sb | 100μg/mL |
Se | 100μg/mL |
Sr | 100μg/mL |
Ti | 100μg/mL |
Tl | 100μg/mL |
V | 100μg/mL |
Zn | 100μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Quality Control Standard 21 | |
Triple-leached LDPE bottle | |
0.01 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Assurance | |
5% HNO3/tr Tart. Acid/tr HF |
Standard 21 | |
250 mL | |
87-69-4,7440-02-0,7439-96-5,7697-37-2,7664-39-3, 108-88-3 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
As,Be,Ca |
Safety and Handling
- quality control standard 21
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : nitric acid
Product intended for laboratory use only