Learn More
Invitrogen™ Quant-iT™ Endotoxin Detection Assay Kit
The Invitrogen Quant-iT Endotoxin Detection Assay Kit is a sensitive and easy-to-use fluorescent endpoint assay for use on fluorescence microplate readers that uses amebocyte lysates to quantify endotoxin in protein, peptide, antibody, or nucleic acid samples.
Brand: Invitrogen™ Q32892
441.35 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
The Invitrogen Quant-iT Endotoxin Detection Assay Kit is a sensitive and easy-to-use fluorescent endpoint assay for use on fluorescence microplate readers that uses amebocyte lysates to quantify endotoxin in protein, peptide, antibody, or nucleic acid samples. Amebocyte lysates are widely used as a simple and sensitive assay for the detection of endotoxin lipopolysaccharide (LPS), the membrane component of gram-negative bacteria.
Features of the Quant-iT Endotoxin Detection Assay Kit include:
• Highly sensitive with a broad range—detect as little as 0.01 EU/mL to 10.0 EU/mL
• Specific—no interference from ß-glucans and suitable for wide range of samples, including protein, vaccine, plasmid, DNA, and RNA
• Easy to use—the simple workflow requires one incubation prior to measurement
Endotoxin levels in the samples are accurately determined using the included endotoxin standard of known concentration that is derived from E. coli strain O111: B4. Determining endotoxin levels is important to assess the efficiency of endotoxin removal methods and prevent endotoxic shock, inflammation, and/or sepsis in tissue culture cells and animals injected with endotoxin-contaminated proteins.
When endotoxin encounters the amebocyte lysate, a series of enzymatic reactions results in the activation of Factor C, Factor B, and pro-clotting enzyme. The activated enzyme catalyzes a cleavage event in the substrate to produce a strong fluorescent signal. After stopping the reaction, the resulting signal is measured on a fluorescence microplate reader (such as Varioskan LUX). The correlation between fluorescent signal is proportional to the endotoxin concentration in the sample.
Notes
• Compatible with Pierce Endotoxin-Free Plate Strips (Cat. No. A43888)
Specifications
Endotoxin | |
Kit | |
160 reactions | |
Protein, Antibody, Plasmid, DNA, RNA |