Learn More
Invitrogen™ Quant-iT™ OliGreen™ ssDNA Assay Kit and Quant-iT OliGreen ssDNA Reagent
Quant-iT™ OliGreen ssDNA Assay Kit and Quant-iT OliGreen ssDNA Reagent contain an ultrasensitive, green-fluorescent nucleic acid stain for quantitating oligonucleotides and single-stranded DNA (ssDNA) in solution.
Brand: Invitrogen™ O7582
Code : 50
Additional Details : Weight : 0.00525kg
Description
Quantify ssDNA with ease using the Quant-iT OliGreen ssDNA Assay kit and reagent. This assay kit and reagent enable sensitive quantitation of single-stranded DNA without the need for a highly concentrated sample. As little as 100 pg/mL can be quantitated using the assay kit or Quant-iT OliGreen ssDNA Reagent.
Quantitate ssDNA and other oligonucleotides accurately and without the need for a highly concentrated sample with Quant-iT OliGreen ssDNA Assay Kit and Quant-iT OliGreen ssDNA Reagent. Quant-iT OliGreen ssDNA Assay Kits contain an ultrasensitive, green-fluorescent nucleic acid stain for quantitating oligonucleotides and single-stranded DNA (ssDNA) in solution. The Quant-iT™ OliGreen™ ssDNA Assay Reagent enables researchers to quantify as little as 1 ng/mL oligonucleotide or ssDNA (200 pg in a 200 μL assay volume) with a fluorescence microplate reader using fluorescein excitation and emission wavelengths. This sensitivity exceeds that achieved with absorbance methods by 10,000-fold.In addition to the dye, the OliGreen ssDNA Assay Kit includes buffer and an oligonucleotide standard. The OliGreen reagent is also available as the stand-alone Quant-iT OliGreen ssDNA Reagent.
Order Info
Shipping Condition: Room temperature
Specifications
Fluorescence | |
Microplate Reader | |
200 pg to 2 μg | |
Quant-iT OliGreen ssDNA Reagent |
500/525 | |
200 assays (2 mL assay volume) | |
OLIGREEN, Quant-iT | |
Room Temperature |