missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuantaBlu™ Fluorogenic Peroxidase Substrate Kit

Conduct stopped, non-stopped and kinetic assays using this soluble fluorogenic (chemifluorescent) peroxidase substrate for ELISA.

Brand:  Thermo Scientific™ 15169

286.64 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 10063933

  • £334.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ QuantaBlu Fluorogenic Peroxidase Substrate is a soluble fluorogenic (chemifluorescent) substrate for detection of peroxidase activity in ELISA methods.

Additional Information and Online Ordering

In the presence of active peroxidase the working solution of QuantaBlu Substrate produces a blue fluorescent product that can be quantitated by fluorometry in microplates or cuvettes. Fluorometric-based detection has a large dynamic range, which overcomes the limitations of using spectrophotometry to measure colorimetric substrates. The blue fluorescent product is not sensitive to light and does not photobleach. QuantaBlu Fluorogenic Peroxidase Substrate is stable, highly sensitive, produces high signal-to-noise ratios and allows for a broad dynamic detection range of peroxidase activity. This substrate is also flexible and can be used for stopped, nonstopped and kinetic assays. Incubation for stopped and nonstopped assays can be performed between 5 and 90 minutes at either room temperature (RT) or 37°C. The QuantaBlu NS/K Substrate formulation can be integrated into robotic-based assays and is designed for assays that do not need to be stopped.

Highlights:

  • More sensitive than TMB, OPD or ABTS substrates
  • Flexible stopped, nonstopped or kinetic assays possible
  • Large dynamic range (4 log peroxidase concentration range)
  • Excellent stability; working solution is stable for 24 hours
  • Large stokes shift; excitation/emission maxima of 325nm/420nm (range of 315-345/370-460)
  • Does not photobleach
TRUSTED_SUSTAINABILITY
Specifications

Specifications

QuantaBlu Fluorogenic Peroxidase Substrate
550 mL
QuantaBlu™
HRP (Horseradish Peroxidase)
ELISA Substrate
Fluorescent
Chemical Substrate
Sufficient For: 275 mL of 1X working solution (2750 wells at 100 μL per well)
• QuantaBlu Substrate Solution, 250 mL
• QuantaBlu Peroxide Solution, 30 mL
• QuantaBlu Stop Solution, 275 mL

Store at 4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK400572 Certificate TypeCertificate of Analysis Date18/12/2024 Catalog Number15169
Lot NumberZI397887 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number15169
Lot NumberZG395932 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number15169
Lot NumberZE393904 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number15169
Lot NumberZC367151 Certificate TypeCertificate of Analysis Date22/03/2024 Catalog Number15169
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ QuantaBlu™ Fluorogenic Peroxidase Substrate Kit > QuantaBlu Peroxidase Substrate with stop solution

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.