Learn More
Thermo Scientific™ Quantech™ Fluorometer Narrow-Band Excitation and Emission Filters
Thermo Scientific™ Quantech Digital Filter Fluorometer Narrow Band Filters allow you to choose the best excitation and emission wavelength ranges for your fluorescence experiment.
Brand:  Thermo Scientific™ LE1095X9
Code : 04
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11701131
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Thermo Scientific™ Quantech Digital Filter Fluorometer Narrow Band Filters allow you to choose the best excitation and emission wavelength ranges for your fluorescence experiment. Narrow band filters allow a narrow range of light to pass through the filter -- delivering the selectivity you need for accurate fluorescence assays.
Narrow band filters can be used to excite fluorescence or to measure a very small window of fluorescence emission. To select the proper filter for your assay, use the following guidelines:
  • There should be no overlap between the Narrow Band excitation filter and the Sharp Cut emission filter
  • A portion of the fluorescence excitation (absorption) spectrum near the peak falls in the wavelength range transmitted by the Narrow Band excitation filter
For the most sensitive measurement of fluorescence, an emission filter that does not overlap with this excitation filter must be selected.
Specifications
Specifications
Narrow Band Filter
420 nm
Quantech