Learn More
Thermo Scientific™ Quantech™ Sharp Cut Fluorometer Filters
Thermo Scientific™ Quantech Sharp Cut Fluorometer Filters deliver accurate results with the sensitivity you demand.
£289.68 - £545.70
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
11721131
|
Thermo Scientific™
LE1095X18 |
415 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10314110
|
Thermo Scientific™
LE1095X19 |
430 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11741131
|
Thermo Scientific™
LE1095X20 |
450 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11751131
|
Thermo Scientific™
LE1095X21 |
475 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11761131
|
Thermo Scientific™
LE1095X22 |
500 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11651369
|
Thermo Scientific™
LE1095X23 |
515 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11771131
|
Thermo Scientific™
LE1095X24 |
535 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11781131
|
Thermo Scientific™
LE1095X25 |
550 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11791131
|
Thermo Scientific™
LE1095X26 |
585 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11701141
|
Thermo Scientific™
LE1095X27 |
605 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11711141
|
Thermo Scientific™
LE1095X29 |
665 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Thermo Scientific™ Quantech Sharp Cut Fluorometer Filters deliver accurate results with the sensitivity you demand.
Sharp cut filters allow all of the light beyond the cut off wavelength to pass through the filter - delivering the maximum amount of signal to the detector for accurate fluorescence assays.Sharp cut filters can be used to measure fluorescence emission and increase detection limits for sensitive measurements.
To select the proper filter for your assay, use the following guidelines:
- There is no overlap between the Narrow Band excitation filter and the Sharp Cut emission filter The majority of the fluorescence emission falls in the wavelength range transmitted by the Sharp Cut emission filter
Includes:
SC415 filter
Note:
Number following SC is wavelength in nanometers. Sharp Cut filters transmit light to the red (lower energy) of the indicated wavelength, and therefore allow the majority of the fluorescence emission to pass through the filter. Should never be used as excitation filters.
Specifications
Sharp Cut Filter | |
Quantech |