Learn More
Thermo Scientific™ Quanti-Cult™ Pseudomonas paraeruginosa ATCC™ 9027™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Brand: Thermo Scientific™ R4735210
Description
Thermo Scientific™ Quanti-Cult™ Pseudomonas paraeruginosa ATCC™ 9027™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial, ready for rehydration and use in quality control procedures.
- No serial dilution required
- Delivers <100 colony forming units (CFUs) in a single 0.3mL inoculum
Ordering Alerts:
Yield is 1 Test/reconstituted vial, 10 Tests/kit
Includes:
10 preserved organism vials and 10 rehydration vials.
Specifications
9027TM* | |
Quanti-Cult | |
Gram-negative Rods | |
10 Test/Kit |
Pseudomonas paraeruginosa ATCC™ 9027™ | |
Bactidisk | |
Quantitative QC |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.