Learn More
R&D Systems™ Quantikine Immunoassay Control Group 235
Controls for Human Adiponectin (HMW), Clusterin, EGF R, and PCSK9
Brand: R&D Systems™ QC235
Additional Details : Weight : 0.00970kg
Description
For use as quantitative controls for the determination of cytokine concentrations in biological fluids. Concentrations have been assigned using R&D Systems™’ Quantikine™ ELISA kits. Controls are prepared in a buffered protein base with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
Specifications
Store the unopened product at 2-8°C. Do not use past expiration date. | |
1 Each |