Learn More
R&D Systems™ Quantikine Immunoassay Control Set 822 Human Total PAI-1
For use with catalog number DTSE100
Brand: R&D Systems™ QC209
Additional Details : Weight : 0.09700kg
Description
For use as quantitative controls for the determination of cytokine concentrations in biological fluids. Concentrations have been assigned using R&D Systems™’ Quantikine™ ELISA kits. Controls are prepared in a buffered protein base with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
Specifications
Store the unopened product at 2-8°C. Do not use past expiration date. | |
5054 | |
1 Each |