Learn More
R&D Systems™ Quantikine Immunoassay Control Set 884 Human FABP2/I-FABP
For use as quantitative controls for the determination of cytokine concentrations in biological fluids
Brand: R&D Systems™ QC213
Additional Details : Weight : 0.09700kg
Description
- Prepared in diluted porcine serum with preservatives
- Contains recombinant human cytokines at low, medium and high concentrations
- Supplied lyophilized
Storage: Unreconstituted Controls should be stored at 2° to 8°C and are stable for at least 6 months from date of receipt. Depending on the analyte of interest, reconstituted controls may be stable when stored at <-20°C. Users should evaluate the frozen stability of the controls in their application or discard after use.
Specifications
189°C | |
FABP2 (I-FABP) | |
2169, 25598 | |
Yes | |
1 Each | |
Unreconstituted Controls are stable for at least 6 months from date of receipt. Depending on the analyte of interest, reconstituted controls may be stable when stored at <-20°C. |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
ELISA Control | |
Control | |
-40°C | |
Human, Rat | |
Liquid |
For Research Use Only.