Learn More
Quantikine Immunoassay Control Set 887 Human TIM-3, R&D Systems™
For use with catalog number DTIM30
Brand: R&D Systems QC217
Additional Details : Weight : 0.09700kg
Description
TIM-3 specifically detects TIM-3 in Human samples. It is validated for Control.Specifications
TIM-3 | |
CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 | |
RUO | |
Reconstitute each vial with the volume of deionized or distilled water indicated on the product datasheet. | |
Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Control | |
1 Each | |
84868 | |
Human |