Learn More
R&D Systems™ Quantikine Immunoassay Control Set 905 for Human NT-ProANP
Immunoassay control set for Human Atrial Natriuretic peptide (ANP) protein
Brand: R&D Systems™ QC227
Additional Details : Weight : 0.48500kg
Description
Atrial Natriuretic peptide (ANP) is synthesized as 151 aa preproprotein. Proprotein contains cardiodilatin (aa 26 - 92) and atrial natriuretic factor (ANF; aa 124 - 151) which become active after processing by serine protease corin. ANP is primarily synthesized in heart and is secreted in response to atrial distention. ANF circulates in plasma, eliciting natriuretic, diuretic, vasorelaxant and antimitogenic effects directed toward reduction of body fluid and blood pressure regulation. ANF acts through cGMP-coupled NP receptor A. Human ANP and ANF share 84% and 96% aa identity with mouse ANP and ANF, respectively.
- Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
4878 (Human); 230899 (Mouse); 24602 (Rat) | |
Immunoassay Control Set 905 | |
Atrial Natriuretic peptide |
Control Set for ELISA Kit | |
For quantitative measurement of Atrial Natriuretic peptide (ANP) levels | |
Human |
For Research Use Only