Learn More
R&D Systems™ Quantikine Immunoassay Control Set 918 for Human IGF-II/IGF2
Immunoassay control set for Human Insulin-like growth factor protein levels.
Brand: R&D Systems™ QC230
Additional Details : Weight : 0.00970kg
Description
Insulin-like growth factor (IGF)-I (also known as somatomedin C and somatomedin A), IGF-II (multiplication stimulating activity or MSA) belongs to family of insulin-like growth factors that are structurally homologous to proinsulin. Mature IGF-I and IGF-II share approximately 70% sequence identity. Both IGF-I and IGF-II are expressed in many tissues, cell types, may have autocrine, paracrine, endocrine functions. Mature IGF-I and IGF-II are highly conserved between human, bovine, porcine proteins (100% identity), exhibits cross-species activity. The IGFL (insulin-like growth factor-like) family includes four small (∼11 kDa), probably secreted family members in humans, one in mouse. This family shares A and B chain cysteine motifs with IGF superfamily, and has additional cysteine motif within uncleaved region corresponding to C peptide of IGF family.
- Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
3481 (Human); 16002 (Mouse) | |
Immunoassay Control Set 930 | |
Insulin-like growth factor (IGF)-I |
Control Set for ELISA Kit | |
For quantitative measurement of Insulin-like Growth Factor II protein levels | |
Human |
For Research Use Only