Learn More
Macherey-Nagel™ Quantofix™ Nitrite Test Strip Kits
Allows for the quick and easy determination of nitrite in solutions. Macherey Nagel™ Quantofix™ Nitrite Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Brand: Macherey-Nagel™ 91311
Description
Allows for the quick and easy determination of nitrite in solutions. Macherey Nagel™ Quantofix™ Nitrite Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
- Ready to use
- Rapid: provides reliable results within one minute
- Excellent for checking cooling lubricants and aquaculture/fish farming
- Precise: colour charts adjusted and check using certified standard solutions
- Shelf life: 2.5 years, store below 30°C
Specifications
Yellow to Red | |
Determination of Nitrite | |
100 Pack | |
1 min. |
Strip | |
0, 1, 5 ,10, 20, 40, 80mg/L NO 2 - | |
30°C | |
Semi-qualitative Test Strip |