missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Quantofix™ Peroxide 25 Semi-quantitative Test Strips

Ideal to test low levels of peroxide in solutions. Macherey Nagel™ Quantofix™ Peroxide 25 Semi-quantitative Test Strips use an easy dip-and-read procedure and provide reliable results within 15 seconds.

Brand:  Macherey-Nagel™ 91319

Product Code. 10528482

  • £61.05 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Ideal to test low levels of peroxide in solutions. Macherey Nagel™ Quantofix™ Peroxide 25 Semi-quantitative Test Strips use an easy dip-and-read procedure and provide reliable results within 15 seconds.

  • Rapid: most tests can be carried out in 10s to 120s
  • Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
  • Precise: color charts adjusted and checked using certified standard solutions
  • Shelf life: 2. 5 years, store below 30°C
Specifications

Specifications

White to Blue
Determination of Peroxide 25
100 Pack
15 sec.
Strip
0, 0.5, 2, 5, 10, 25mg/L H2O2
30°C
Semi-qualitative Test Strip
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Quantofix™ Peroxide 25 Semi-quantitative Test Strips > Color Indicator: White to Blue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.