Learn More
Macherey-Nagel™ Quantofix™ Sulfate Test Strip Kits
Allows for the quick and easy determination of sulfate in solutions. Macherey Nagel™ Quantofix™ Sulfate Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Brand: Macherey-Nagel™ 91329
This item is not returnable.
View return policy
Description
- Rapid: provides reliable results within two minutes
- Excellent for use in the beverage industry
- Precise: colour charts adjusted and checked using certified standard solutions
- Shelf life: 2.5 years, store below 30°C
Specifications
Red to Yellow | |
Determination of Sulfate | |
100 Pack | |
2 min. |
Strip | |
<200, >400, >800, >1200, >1600mg/L SO42- | |
30°C | |
Semi-qualitative Test Strip |