Learn More
Applied Biosystems™ QuantStudio™ 3D Digital PCR Chip Adapter Kit for Flat Block Thermal Cycler
Required accessory for securely positioning.
Brand: Applied Biosystems™ 4485513
Additional Details : Weight : 0.09200kg
Description
QuantStudio™ 3D Digital PCR Chips in the dual flat blocks of the GeneAmp™ 9700 or ProFlex™ thermal cyclers. The Adaptor Kit includes two block adaptors, as well as two compression pads that sit atop the chips during thermal cycling.
Specifications
Contents: • 2 chip adapters • 2 thermal pads |
|
Digital PCR | |
QuantStudio™ | |
Real Time PCR (qPCR) |
3D Digital PCR Chips | |
GeneAmp 9700, GeneAmp™ PCR System 9700, Dual Flat Block, ProFlex™ 2x Flat PCR System, ProFlex™ PCR System | |
3d Digital PCR Chip Adapter Kit |