missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ QuantStudio 7 PRO 0.2mL, 96-well package
The Applied Biosystems QuantStudio 7 Pro Real-Time PCR System adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint. Offers facial authentication, voice commands, Smart Help, multiplex capability, motorized interchangeable blocks, simplified workflow, intuitive software, and large capacitive touch-screen interface.
Specifications
Thermal Uniformity | 0.4°C |
---|---|
Capacity | 96 |
Display Type | Touchscreen |
Height (Metric) | 54.7 cm |
Width (Metric) | 33.8 cm |
Product Code | Brand | Content And Storage | Warranty | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Content And Storage | Warranty | Price | Quantity & Availability | ||||
16277631
|
Applied Biosystems™
A44289 |
No PC | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
N/A
|
N/A | ||||
16297631
|
Applied Biosystems™
A44291 |
PC (Laptop) | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
N/A
|
N/A | ||||
16217641
|
Applied Biosystems™
A44293 |
PC (Tower) | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
N/A
|
N/A | ||||
Description
Features of the QuantStudio 7 Pro Real-Time PCR System include:
- Easy and convenient hands-free operation with facial authentication and voice commands
- Reduced errors and manual entries with smart qPCR workflow and TaqMan Array Plates with RFID
- Space savings with small footprint and true standalone operation; set-up, start, monitor, and analyze directly on the touchscreen
- Peace of mind with Smart Help; contact Technical Support directly from the touchscreen
- Reduced downtime with Smart Remote Support; show issues to Technical Support for speedy resolution
- Application versatility with easily changed blocks and four different block types
- Increased productivity with automation compatibility for 24/7 operation
- Access, analyze, and share data anytime, anywhere with our Connect cloud-based platform for real-time information access, secure data storage, and online results sharing
- Remotely monitor experiments using the Instrument Connect monitoring app
- Design and Analysis software compatible with PC and Mac operating systems
- Compatible with over 8 million TaqMan Assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- Provides exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation
- Designed for ultimate ease-of-use, the system offers high quality, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab
Specifications
0.4°C | |
Touchscreen | |
33.8 cm | |
52.5 cm | |
6.5°C/s | |
Bright White LED | |
5/6 Targets | |
FAM™, SYBR™ Green, VIC™, JOE™, HEX™, TET™, ABY™, NED™, TAMRA™, Cy™3, JUN™, ROX™, Texas Red™, Mustang Purple™, Cy™5, LIZ™, Cy™5.5 | |
96-well |
96 | |
54.7 cm | |
1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) | |
4°C to 99.9°C | |
QuantStudio 7 PRO | |
0.2 mL (96-well) | |
CMOS Technology | |
38 kg |