missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ QuantStudio™ 7 Pro Real-Time PCR System Extended Warranty Package, 96-well, 0.2 mL
The Applied Biosystems QuantStudio 7 Pro Real-Time PCR System adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint. Package includes the QuantStudio 7 Pro Real-Time PCR System, SmartSmart Orientation, and a 1 or 3 year extended warranty.
£85620.00
Specifications
Block Format | Interchangeable |
---|---|
For Use With (Equipment) | Real Time Thermal Cyclers |
Display Type | 12 in. Color Touchscreen |
Green Features | Energy Efficient, Fewer Resources used and Less Waste |
Shipping Condition | Room Temperature |
Description
- Adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint
- The QuantStudio 7 Pro system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation
- Designed for ultimate ease-of-use, the system offers high quality, high throughput, and excellent reliability
- Easy and convenient hands-free operation with facial authentication and voice commands
- Reduced errors and manual entries with smart qPCR workflow and TaqMan Array Plates with RFID
- Space savings with small footprint and true standalone operation; setup, start, monitor, and analyze directly on the touchscreen
- Compatible with over 8 million TaqMan Assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- An optimal user experience for researchers who want to work smarter in the lab
- Package includes the QuantStudio 7 Pro Real-Time PCR System, SmartSmart Orientation, and a 1 or 3 year extended warranty
- Extended warranty includes comprehensive service coverage and one planned maintenance visit
Specifications
Interchangeable | |
12 in. Color Touchscreen | |
Room Temperature | |
1 Year Manufacturer Warranty and 1 Year of Field Service with PM |
Real Time Thermal Cyclers | |
Energy Efficient, Fewer Resources used and Less Waste | |
1 Day at Customer Site |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Applied Biosystems™ QuantStudio™ 7 Pro Real-Time PCR System Extended Warranty Package, 96-well, 0.2 mL