missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ Absolute Q™ Isolation Buffer

The QuantStudio Absolute Q Isolation Buffer is intended to supplement the materials supplied in the QuantStudio Absolute Q MAP16 Plate Kit.

Brand:  Applied Biosystems™ a52730

Product Code. 17427713

  • £64.50 / 3mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The QuantStudio Absolute Q Isolation Buffer is one component of the QuantStudio Absolute Q MAP16 Plate Kit (A52865)
  • This buffer is intended to supplement the materials supplied in the plate kit for those customers who require additional materials
  • For use within the QuantStudio Absolute Q Digital PCR System workflow to help partition samples within the reaction wells of the MAP16 plate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Primer-probe
QuantStudio™
Room Temperature
QuantStudio Absolute Q MAP16 Plate Kit
QuantStudio™ Absolute Q™ Digital PCR System
3 mL
Digital PCR (dPCR)
3 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2835799 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberA52730
Lot Number2865093 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberA52730
Lot Number2914645 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberA52730
Lot Number2936053 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA52730
Lot Number2966505 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA52730
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ QuantStudio™ Absolute Q™ Isolation Buffer >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures