Learn More
Bellingham Stanley™ Quartz Control Plate
High quality quartz control plates for verification of polarimeters
Brand:  Bellingham Stanley™ 34-21
Product Code. 11900108
  • £1625.00 / Each
Estimated Shipment: 03-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Bellingham and Stanley offer a number of traceable calibration materials for verifying and calibrating refractometers. AG fluids and calibration oils are supplied in packs of 5 x 5mL glass bottles and accompanied by a UKAS Certificate of Calibration showing values at 20°C and 589.3nm, these certified reference materials provide an ideal solution to your refractometer calibration needs. Quartz Control Plates (QCP) are also offered for verifying and calibrating polarimeters and are supplied with a Certificate of Conformity showing values at sodium and NIR for both angular and sugar scales. A thermal block is available for use with Bellingham and Stanley ADP polarimeters, it houses the external temperature sensor enabling the use of quartz temperature compensation. QCPs can be supplied with UKAS Certificate of Calibration showing traceability to PTB as an optional service.
For temperatures other than 20°C, both RI and Brix values are available from the manufacturer's website.
RI and Brix values
  • Wide choice of values
  • 12 month validity
  • UKAS Certificate of Calibration
  • Traceable to NIST and ICUMSA
Thermal block for use with ADP/S available, please enquire.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Quartz Control Plate
Quartz
5.2 A
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Bellingham Stanley™ Quartz Control Plate >