Learn More
Thermo Scientific Chemicals Quartz, cover slip for microscope slide, fused, 25.4x25.4x0.15-0.25mm (1.0x1.0x0.006-0.01in)
For use with microscope quartz slides made by fusing naturally occurring crystalline silica
Brand: Thermo Scientific Chemicals 043210.KG
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Cover slips for use with microscope quartz slides
Specifications
0.15 to 0.25 mm | |
Cover Slips for use with Microscope Quartz Slide | |
1/Ea. | |
25.4 x 25.4 mm |
RUO – Research Use Only