Learn More
Bel-Art™ SP Scienceware™ Wilmad™ X-Band Capillary EPR Tubes for Aqueous Sample
Close up the end with a hot flame or with beeswax. Bel Art SP Scienceware™ Wilmad™ X-Band Capillary EPR Tube for Aqueous Sample are often used in EPR applications that require tuning of aqueous samples at high dB or where sample size is limited.
Brand:  Bel-Art™ 66000-0040
Product Code. 15348608
  • £235.00 / Pack of 25
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Open on both ends
  • Easy to seal with a hot flame or beeswax
Specifications
Specifications
100 mm
1.0 mm
Q-1.0X1.2
1.2 mm
25 Pack