Learn More
Invitrogen™ Qubit™ 1X dsDNA High Sensitivity (HS) and Broad Range (BR) Assay Kits

Description
Experience easy, accurate, and precise quantification of dsDNA with Qubit 1X dsDNA HS (High Sensitivity) and Qubit 1X dsDNA BR (Broad Range) Assay Kits. The 1X formulation provides a ready-to-use working solution that enables simple dsDNA sample quantification on Qubit Fluorometers. These assays are highly selective for dsDNA over ssDNA, RNA, protein, and free nucleotides. Contaminants, such as salts, solvents, or detergents are well-tolerated.
Qubit 1X dsDNA HS Assay Kit
The Qubit dsDNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of sensitive DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 5 pg/μL to 120 ng/μL, providing a detection range of 0.1−120 ng.
Qubit 1X dsDNA BR Assay Kit
The Qubit dsDNA BR (Broad-Range) Assay Kit, designed for use with Qubit 4 or Qubit Flex Fluorometers, provides an accurate and selective method for the quantitation of DNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 0.2 to 4,000 ng/μL, providing a detection range of 4-4,000 ng.
Notes
• Qubit 1X dsDNA HS can be used with Qubit 2, Qubit 3, Qubit 4, and Qubit Flex Fluorometers
• Qubit 1X dsDNA BR are designed for use with Qubit 4 and Qubit Flex Fluorometers
• Use thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200 μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| Assay | 1X dsDNA, broad range |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Qubit 4 and Qubit Flex Fluorometer |
| Green Features | Fewer resources used, Less waste |
| No. of Reactions | 100 Reactions |
| Quantitation Range | 4 to 4000 ng |
| Product Line | Qubit |
| Quantity | 100 assays |
| Shipping Condition | This assay kit will ship as two components. • Qubit™ 1X dsDNA BR Working Solution will be shipped (at ambient conditions.) • Qubit 1X dsDNA BR lambda standards will be shipped on wet ice. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.