Learn More
Invitrogen™ Qubit™ 4 Fluorometer, with WiFi
Qubit™ 4 Flurometer with WiFi is designed to accurately measure DNA, RNA, and protein quantity, and now also RNA integrity and quality. Re-engineered to enable data transfer via WiFi as well as to run the Qubit™ RNA IQ (integrity & quality) assay.
Brand: Invitrogen™ Q33238
3070.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Qubit™ 4 Fluorometer and RNA IQ Assay Kit work together to accurately distinguish intact from degraded RNA in just two easy steps
- For all assays, the concentration or quality of the target molecule in the sample is reported by a fluorescent dye that emits a signal only when bound to the target, which minimizes the effects of contaminants on the result
- Easy-to-use touch-screen menus make it easy to select and run the assays, with results displayed in just a few seconds
Features:
- Fast and highly accurate quantitation of DNA, RNA, and protein in less than three seconds per sample
- Measures intact RNA in less than 5 seconds per sample
- High levels of accuracy using only 1 to 20 μL of sample, even with very dilute samples
- On-board Reagent Calculator quickly generates Qubit™ working solution preparation instructions
- Stores results from up to 1000 samples
- Large 5.7 in, state-of-the-art, color touch screen for easy workflow navigation
- Graphical display indicates when samples are in the extended range or out of range
- Small footprint saves space on your bench
- Flexibility to export data to using the included WiFi dongle, a USB drive, or directly to your computer via a USB cable
- Ability to personalize your Qubit™ fluorometer with the assays you run most, add new assays, or even create your own assays with the MyQubit™ software and web tool
- Language of your choice including English, French, Spanish, Italian, German, simplified Chinese, and Japanese
- Pre-loaded program to test performance of fluorometer using the Qubit™ Fluorometer Verification Assay Kit
Flurometer can also be used to directly measure the fluorescence of samples using Fluorometer mode.
- Ion Sphere Particle quality can be evaluated on the Fluorometer using the Ion Sphere Quality Control Kit prior to performing a sequencing run on the Ion PGM sequencer.
Enables more accurate results than with UV absorbance at low concentrations
- Fluorometer is far more sensitive than UV absorbance, which measures anything absorbing at 260 nm, including DNA, RNA, protein, free nucleotides, or excess salts
- Dyes in the Qubit™ assay kits fluoresce only when bound to the selected molecule (DNA, RNA or protein) in a sample, preventing repetating work due to inaccurate measurements
Fast, simple, and accurate measurement of RNA quality
- Qubit™ RNA IQ assay provides a fast, simple method to check whether an RNA sample has degraded
- Assay utilizes two unique dyes: one that binds to large, intact RNA, and another that selectively binds to small, degraded RNA, together enabling quick assesment of the quality and integrity of an RNA sample.
- To use, simply add samples to the RNA IQ working solution, then measure on the Fluorometer.
Save samples and bench space
- Requires samples of only 1 to 20 μL
- Intended for use at room temperature and occupies only a small amount of bench space
- Features advanced optics and data analysis algorithms, and comes equipped with a WiFi dongle, a USB thumb drive, and cable for data transfer to Excel software and for software downloads, as well as a universal power supply, four plug adapters, and electromagnetic CE certification
- Enables maximum flexibility for data export using WiFi, USB drive, or USB cable direct to computer.
- All data can be saved in the cloud application, including direct transfer from the instrument via WiFi.
Single sample measurements are a breeze
- User-friendly fluorometer is combined with quick and simple assay procedures
- Calculations and settings are automatically performed by the instrument
- Asays are all performed using the same general protocol, which uses a simple mix-and-read format with incubation times of only two minutes for the DNA and RNA assays.
- 500 L thin-walled PCR tubes are required

Specifications
1 to 20 μL | |
Qubit Fluorometer | |
Fluorometer | |
Room Temperature |
470 nm (Blue LED), 635 nm (Red LED) | |
Qubit | |
DNA Quantification, RNA Quantification, Protein Quantification | |
1 fluorometer |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.